N-Methyl-1-(4-morpholin-4-ylphenyl)ethanamine

Supplier: Matrix Scientific

064626-500MG
101928-062EA 727.69 USD
101928-062
N-Methyl-1-(4-morpholin-4-ylphenyl)ethanamine
N-Methyl-1-(4-morpholin-4-ylphenyl)ethanamine

Hazard: Irritant

Formula: C₁₃H₂₀N₂O
MDL Number: MFCD08060585
CAS Number: 920483-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR