2-Cyclopropylbutan-2-ol

Supplier: Matrix Scientific

064605-500MG
101928-018EA 484.62 USD
101928-018
2-Cyclopropylbutan-2-ol
2-Cyclopropylbutan-2-ol

Hazard: Irritant

Formula: C₇H₁₄O
MDL Number: MFCD00182411
CAS Number: 4435-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR