4-Bromo-3-ethyl-5-methyl-1H-pyrazole

Supplier: Matrix Scientific

064456-500MG
101927-738EA 484.62 USD
101927-738
4-Bromo-3-ethyl-5-methyl-1H-pyrazole
4-Bromo-3-ethyl-5-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₆H₉BrN₂
MDL Number: MFCD17078878
CAS Number: 736073-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR