3-(3,5-Dimethyl-1H-1,2,4-triazol-1-yl)propanenitrile

Supplier: Matrix Scientific

064363-500MG
101927-552EA 484.62 USD
101927-552
3-(3,5-Dimethyl-1H-1,2,4-triazol-1-yl)propanenitrile
3-(3,5-Dimethyl-1H-1,2,4-triazol-1-yl)propanenitrile

Hazard: Irritant

Formula: C₇H₁₀N₄
MW: 150.18 g/mol
MDL Number: MFCD14630742
CAS Number: 60866-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR