2-[2-(1H-Pyrazol-4-yl)ethyl]-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

063992-500MG
101926-782EA 364.62 USD
101926-782
2-[2-(1H-Pyrazol-4-yl)ethyl]-1H-isoindole-1,3(2H)-dione
2-[2-(1H-Pyrazol-4-yl)ethyl]-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₃H₁₁N₃O₂
MDL Number: MFCD16618465
CAS Number: 1195687-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR