6-Benzyl-2-mercaptopyrimidin-4(3H)-one

Supplier: Matrix Scientific

Synonyms: 6-Benzyl-2-thioxo-2,3-dihydropyrimidin-4(1H)-one, Benzylthiouracil

063884-500MG
101926-568EA 364.62 USD
101926-568
6-Benzyl-2-mercaptopyrimidin-4(3H)-one
6-Benzyl-2-mercaptopyrimidin-4(3H)-one

Hazard: Irritant

Formula: C₁₁H₁₀N₂OS
MDL Number: MFCD16618433
CAS Number: 6336-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR