2-Methyl-2-adamantanol

Supplier: Matrix Scientific

Synonyms: 2-Hydroxy-2-methyladamantane, 2-Methyladamantan-2-ol

063692-500MG
101926-212EA 146.15 USD
101926-212
2-Methyl-2-adamantanol
2-Methyl-2-adamantanol

Hazard: Irritant

Formula: C₁₁H₁₈O
MW: 166.26 g/mol
MDL Number: MFCD00074811
CAS Number: 702-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR