2-Adamantanone oxime

Supplier: Matrix Scientific

063651-500MG
101926-134EA 183.08 USD
101926-134
2-Adamantanone oxime
2-Adamantanone oxime

Hazard: Irritant

Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
Melting Pt: 162…163 °C
MDL Number: MFCD00078273
CAS Number: 4500-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR