3-[(3-Bromophenyl)amino]cyclohex-2-en-1-one

Supplier: Matrix Scientific

063551-500MG
101925-940EA 484.62 USD
101925-940
3-[(3-Bromophenyl)amino]cyclohex-2-en-1-one
3-[(3-Bromophenyl)amino]cyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₂H₁₂BrNO
MDL Number: MFCD15146482
CAS Number: 1217862-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR