5,5-Dimethyl-3-[(3-nitrophenyl)amino]cyclohex-2-en-1-one

Supplier: Matrix Scientific

063522-500MG
101925-882EA 484.62 USD
101925-882
5,5-Dimethyl-3-[(3-nitrophenyl)amino]cyclohex-2-en-1-one
5,5-Dimethyl-3-[(3-nitrophenyl)amino]cyclohex-2-en-1-one

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₃
MDL Number: MFCD00129242
CAS Number: 61997-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR