N-Methyl-2-nitroaniline for synthesis

Supplier: Matrix Scientific

063420-500MG
101925-678EA 183.08 USD
101925-678
N-Methyl-2-nitroaniline for synthesis
N-Methyl-2-nitroaniline
Formula: C₇H₈N₂O₂
Boiling Pt: 158 °C (18 mmHg)
Melting Pt: 38 °C
MDL Number: MFCD00007090
CAS Number: 612-28-2
EINECS: 210-303-1
UN: 2811
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR