6-(Dimethylamino)nicotinic acid

Supplier: Matrix Scientific

Synonyms: 2-(N,N-Dimethylamino)pyridine-5-carboxylic acid

063392-500MG
101925-620EA 484.62 USD
101925-620
6-(Dimethylamino)nicotinic acid
6-(Dimethylamino)nicotinic acid

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD07357408
CAS Number: 82846-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR