N-[Amino(4,4-dimethyl-2,6-dioxocyclohexylidene)methyl]benzamide

Supplier: Matrix Scientific

063170-500MG
101925-278EA 364.62 USD
101925-278
N-[Amino(4,4-dimethyl-2,6-dioxocyclohexylidene)methyl]benzamide
N-[Amino(4,4-dimethyl-2,6-dioxocyclohexylidene)methyl]benzamide

Hazard: Irritant

Formula: C₁₆H₁₈N₂O₃
MDL Number: MFCD00811692
CAS Number: 123865-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR