N-[(4-Methylphenyl)sulfonyl]-beta-alanine

Supplier: Matrix Scientific

Synonyms: 3-(4-Methylphenylsulfonamido)propanoic acid, 3-(4-Methylphenylsulphonamido)propanoic acid

063090-500MG
101925-122EA 243.08 USD
101925-122
N-[(4-Methylphenyl)sulfonyl]-beta-alanine
N-[(4-Methylphenyl)sulfonyl]-beta-alanine

Hazard: Irritant

Formula: C₁₀H₁₃NO₄S
MDL Number: MFCD00453693
CAS Number: 42908-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR