1-Methyltetrahydropyrimidin-2(1H)-one

Supplier: Matrix Scientific

063019-500MG
101924-992EA 330.77 USD
101924-992
1-Methyltetrahydropyrimidin-2(1H)-one
1-Methyltetrahydropyrimidin-2(1H)-one

Hazard: Irritant

Formula: C₅H₁₀N₂O
MDL Number: MFCD00196814
CAS Number: 10166-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR