4-{[(1,1-Dioxido-1,2-benzisothiazol-3-yl)(4-methylphenyl)amino]methyl}-2-methoxyphenol

Supplier: Matrix Scientific

062950-500MG
101924-864EA 243.08 USD
101924-864
4-{[(1,1-Dioxido-1,2-benzisothiazol-3-yl)(4-methylphenyl)amino]methyl}-2-methoxyphenol
4-{[(1,1-Dioxido-1,2-benzisothiazol-3-yl)(4-methylphenyl)amino]methyl}-2-methoxyphenol

Hazard: Irritant

Formula: C₂₂H₂₀N₂O₄S
MDL Number: MFCD03270598
CAS Number: 433941-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR