6-Bromobenzo[d]thiazol-2(3H)-one

Supplier: Matrix Scientific

Synonyms: 6-Bromo-2-benzothiazolinone

062891-5G 062891-25G
101924-746EA 40 USD
101924-746 102551-594
6-Bromobenzo[d]thiazol-2(3H)-one
6-Bromobenzo[d]thiazol-2(3H)-one

Hazard: Irritant

Formula: C₇H₄BrNOS
MDL Number: MFCD00239363
CAS Number: 62266-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR