3,5-Bis(difluoromethyl)-1H-pyrazole

Supplier: Matrix Scientific

062803-500MG
101924-580EA 541.54 USD
101924-580
3,5-Bis(difluoromethyl)-1H-pyrazole
3,5-Bis(difluoromethyl)-1H-pyrazole

Hazard: Irritant

Formula: C₅H₄F₄N₂
MDL Number: MFCD03419821
CAS Number: 77614-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR