N-Methyl-1-(tetrahydro-2H-pyran-2-yl)methanamine

Supplier: Matrix Scientific

062700-500MG
101924-386EA 484.62 USD
101924-386
N-Methyl-1-(tetrahydro-2H-pyran-2-yl)methanamine
N-Methyl-1-(tetrahydro-2H-pyran-2-yl)methanamine

Hazard: Irritant

Formula: C₇H₁₅NO
MW: 129.2 g/mol
MDL Number: MFCD06738978
CAS Number: 7179-96-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR