2-(3,4,5-Trimethoxyphenyl)quinoline-4-carbonyl chloride

Supplier: Matrix Scientific

062283-500MG
101923-658EA 484.62 USD
101923-658
2-(3,4,5-Trimethoxyphenyl)quinoline-4-carbonyl chloride
2-(3,4,5-Trimethoxyphenyl)quinoline-4-carbonyl chloride

Hazard: Irritant

Formula: C₁₉H₁₆ClNO₄
MW: 357.8 g/mol
MDL Number: MFCD03421079
CAS Number: 1160260-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR