6-Chloro-2-(4-isopropylphenyl)quinoline-4-carbonyl chloride

Supplier: Matrix Scientific

061680-500MG
101922-452EA 364.62 USD
101922-452
6-Chloro-2-(4-isopropylphenyl)quinoline-4-carbonyl chloride
6-Chloro-2-(4-isopropylphenyl)quinoline-4-carbonyl chloride

Hazard: Irritant

Formula: C₁₉H₁₅Cl₂NO
MDL Number: MFCD03421291
CAS Number: 1160263-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR