2-(Biphenyl-4-yloxy)propanoyl chloride

Supplier: Matrix Scientific

061486-500MG
101922-064EA 364.62 USD
101922-064
2-(Biphenyl-4-yloxy)propanoyl chloride
2-(Biphenyl-4-yloxy)propanoyl chloride

Hazard: Irritant

Formula: C₁₅H₁₃ClO₂
MDL Number: MFCD12197773
CAS Number: 1160264-64-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR