[2-(Pyridin-3-yloxy)propyl]amine

Supplier: Matrix Scientific

061454-500MG
101921-998EA 484.62 USD
101921-998
[2-(Pyridin-3-yloxy)propyl]amine
[2-(Pyridin-3-yloxy)propyl]amine

Hazard: Irritant

Formula: C₈H₁₂N₂O
MDL Number: MFCD03840189
CAS Number: 886763-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR