2,2'-Ethane-1,2-diylbis-1H-benzimidazole

Supplier: Matrix Scientific

061304-500MG
101921-706EA 243.08 USD
101921-706
2,2'-Ethane-1,2-diylbis-1H-benzimidazole
2,2'-Ethane-1,2-diylbis-1H-benzimidazole

Hazard: Irritant

Formula: C₁₆H₁₄N₄
Melting Pt: 360 °C
MDL Number: MFCD00087492
CAS Number: 3575-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR