5-(1-Naphthylamino)-5-oxopentanoic acid

Supplier: Matrix Scientific

061300-500MG
101921-698EA 146.15 USD
101921-698
5-(1-Naphthylamino)-5-oxopentanoic acid
5-(1-Naphthylamino)-5-oxopentanoic acid

Hazard: Irritant

Formula: C₁₅H₁₅NO₃
MDL Number: MFCD00033064
CAS Number: 296275-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR