1-(1-Naphthylmethyl)-1H-indole-2,3-dione

Supplier: Matrix Scientific

061125-500MG
101921-350EA 243.08 USD
101921-350
1-(1-Naphthylmethyl)-1H-indole-2,3-dione
1-(1-Naphthylmethyl)-1H-indole-2,3-dione

Hazard: Irritant

Formula: C₁₉H₁₃NO₂
MW: 287.32 g/mol
MDL Number: MFCD00498556
CAS Number: 79183-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR