Methyl N-(3,4,5,6-tetrahydro-2H-azepin-7-yl)glycinate hydrochloride

Supplier: Matrix Scientific

061046-500MG
101921-194EA 484.62 USD
101921-194
Methyl N-(3,4,5,6-tetrahydro-2H-azepin-7-yl)glycinate hydrochloride
Methyl N-(3,4,5,6-tetrahydro-2H-azepin-7-yl)glycinate hydrochloride

Hazard: Irritant

Formula: C₉H₁₇ClN₂O₂
MW: 220.7 g/mol
MDL Number: MFCD19103573
CAS Number: 1332530-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR