6-Chloro-3-methoxy-2,3-dihydroimidazo[1,2-b]pyridazine hydrochloride

Supplier: Matrix Scientific

061041-500MG
101921-184EA 606.15 USD
101921-184
6-Chloro-3-methoxy-2,3-dihydroimidazo[1,2-b]pyridazine hydrochloride
6-Chloro-3-methoxy-2,3-dihydroimidazo[1,2-b]pyridazine hydrochloride

Hazard: Irritant

Formula: C₇H₉Cl₂N₃O
MDL Number: MFCD19103506
CAS Number: 1332530-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR