N-Methyl-1-(1,4,5-trimethyl-1H-imidazol-2-yl)methanamine dihydrochloride

Supplier: Matrix Scientific

060812-500MG
101920-732EA 727.69 USD
101920-732
N-Methyl-1-(1,4,5-trimethyl-1H-imidazol-2-yl)methanamine dihydrochloride
N-Methyl-1-(1,4,5-trimethyl-1H-imidazol-2-yl)methanamine dihydrochloride

Hazard: Irritant

Formula: C₈H₁₇Cl₂N₃
MDL Number: MFCD18071329
CAS Number: 1332531-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR