[1-Methyl-2-(1,3-thiazol-2-yl)ethyl]amine dihydrochloride

Supplier: Matrix Scientific

060810-500MG
101920-728EA 969.23 USD
101920-728
[1-Methyl-2-(1,3-thiazol-2-yl)ethyl]amine dihydrochloride
[1-Methyl-2-(1,3-thiazol-2-yl)ethyl]amine dihydrochloride

Hazard: Irritant

Formula: C₆H₁₂Cl₂N₂S
MDL Number: MFCD18071416
CAS Number: 92932-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR