DMS (Dimethyl suberimidate dihydrochloride)
Supplier: Matrix Scientific
060537-500MG
101920-196EA
364.62
USD
101920-196
DMS (Dimethyl suberimidate dihydrochloride)
DMS (Dimethyl suberimidate dihydrochloride)
CAS: 34490-86-3
MDL Number: MFCD00012574
Molecular Formula: C10H22Cl2N2O2
Molecular Weight: 273,21
Hazard: Irritant
Formula:
C₁₀H₂₀N₂O₂ |
MDL Number:
MFCD00012574 CAS Number: 34490-86-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...