IUD Extractor, OR Grade, Sklar
Supplier: Sklar
90-8210
10192-330EA 138.08 USD
10192-330
IUD Extractor, OR Grade, Sklar
Dissection Needles
Sklar's® IUD Extractor is an OB/GYN instrument used for removal of an intrauterine device.
  • Reusable
  • Non-sterile
  • Latex-free
This is a narrow instrument with a slight angle in the shaft and two crochet style hooks on each side of the tip used to grasp the IUD strings.