ReliaPrep™ Non-sterile Glass Pre-filter Syringe Filters, Ahlstrom
Supplier: Ahlstrom
760529
10192-196PK 1373.64 USD
10192-196
ReliaPrep™ Non-sterile Glass Pre-filter Syringe Filters, Ahlstrom
Filters Syringe Filters
Ahlstrom offers syringe filters for convenient filtration of small sample sizes.
This microfiber glass prefilter can be used to remove particles from solution that might block a membrane filter.