ReliaDisc™ Nonsterile Plain Membranes, Ahlstrom
Supplier: Ahlstrom
760181
10192-172PK 160.94 USD
10192-172
ReliaDisc™ Nonsterile Plain Membranes, Ahlstrom
Filters Membrane Filters
Hydrophilic membranes for use as surface micro-filtration filters.
Nylon (polyamide) membrane is useful in both aqueous and organic solvent fitlration applications. Cellulose acetate membrane is low protein binding and provides chemical resistance to alcohol and oil.