ReliaPrep™ Non sterile Cellulose Acetate Syringe Filters, Ahlstrom
Supplier: Ahlstrom
760507
10192-156PK 974 USD
10192-156 10192-158
ReliaPrep™ Non sterile Cellulose Acetate Syringe Filters, Ahlstrom
Filters Syringe Filters
Ahlstrom offers syringe filters for convenient filtration of small sample sizes through membranes.
Cellulose acetate membrane is low protein binding and provides chemical resistance to alcohol and oil.