Sodium hypochlorite 5 - 6% USP

Supplier: HAWKINS PC
Danger

36211
10192-152CS 251 USD
10192-152
Sodium hypochlorite 5 - 6% USP
Sodium hypochlorite
Formula: ClNaO
MW: 74.44 g/mol
Melting Pt: -6
MDL Number: MFCD00011120
CAS Number: 7681-52-9
UN: 1791
ADR: 8 III
Merck Index: 13,08702

Order Now

Specification Test Results

Identification A, IR 197 Conforms
Identification A* Conforms
Identification B* Conforms
Assay, wt. % 5.0 - 6.0 %
Appearance Clear to light yellow
Phosphate, P04, ppm* ≤5 ppm
Calcium, Ca, ppm* ≤10 ppm
*Tested on periodic lots


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR