[4-(2-Oxo-2-pyrrolidin-1-ylethyl)phenyl]amine hydrochloride

Supplier: Matrix Scientific

060181-500MG
101919-566EA 183.08 USD
101919-566
[4-(2-Oxo-2-pyrrolidin-1-ylethyl)phenyl]amine hydrochloride
[4-(2-Oxo-2-pyrrolidin-1-ylethyl)phenyl]amine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O
MDL Number: MFCD11844252
CAS Number: 926265-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR