Ethyl-2-(5-chloro-1,3-benzoxazol-2-yl)acetate ≥95%

Supplier: Matrix Scientific

059646-500MG 059646-1G 059646-5G
101918-744EA 355.38 USD
101918-744 101918-746 101918-748
Ethyl-2-(5-chloro-1,3-benzoxazol-2-yl)acetate ≥95%
Ethyl-2-(5-chloro-1,3-benzoxazol-2-yl)acetate

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₃
Melting Pt: 91…92 °C
MDL Number: MFCD04210543
CAS Number: 138420-09-4

Order Now

Specification Test Results

Melting point 91-92°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR