Ethyl-2-(5-chloro-1,3-benzoxazol-2-yl)acetate ≥95%
Supplier: Matrix Scientific
059646-500MG
059646-1G
059646-5G
101918-744EA
355.38
USD
101918-744
101918-746
101918-748
Ethyl-2-(5-chloro-1,3-benzoxazol-2-yl)acetate ≥95%
Ethyl-2-(5-chloro-1,3-benzoxazol-2-yl)acetate
Hazard: Irritant
Formula:
C₁₁H₁₀ClNO₃ Melting Pt: 91…92 °C |
MDL Number:
MFCD04210543 CAS Number: 138420-09-4 |
Specification Test Results
Melting point | 91-92°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...