1-{2-[(4,6-Dichloro-2-methylpyrimidin-5-yl)amino]-4,5-dihydro-1H-imidazol-1-yl}ethan-1-one ≥95%

Supplier: Matrix Scientific

059637-500MG 059637-1G 059637-5G
101918-706EA 373.85 USD
101918-706 101918-708 101918-710
1-{2-[(4,6-Dichloro-2-methylpyrimidin-5-yl)amino]-4,5-dihydro-1H-imidazol-1-yl}ethan-1-one ≥95%
1-{2-[(4,6-Dichloro-2-methylpyrimidin-5-yl)amino]-4,5-dihydro-1H-imidazol-1-yl}ethan-1-one

Hazard: Irritant

Formula: C₁₀H₁₁Cl₂N₅O
Melting Pt: 246 °C
MDL Number: MFCD04117980
CAS Number: 75438-54-9

Order Now

Specification Test Results

Melting point 246°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR