1-Bromo-4-[(trifluoromethyl)sulfonyl]benzene ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Bromo-4-((trifluoromethyl)sulphonyl)benzene, 1-Bromo-4-((trifluoromethyl)sulfonyl)benzene

059617-1G
101918-654EA 243.08 USD
101918-654
1-Bromo-4-[(trifluoromethyl)sulfonyl]benzene ≥97%
1-Bromo-4-[(trifluoromethyl)sulfonyl]benzene

Hazard: Irritant

Formula: C₇H₄BrF₃O₂S
Melting Pt: 60…65 °C
MDL Number: MFCD00474580
CAS Number: 312-20-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR