2-(2-Chloro-5-nitrophenyl)acetonitrile ≥95%

Supplier: Matrix Scientific

059294-500MG 059294-1G
101917-796EA 598.46 USD
101917-796 101917-798
2-(2-Chloro-5-nitrophenyl)acetonitrile ≥95%
2-(2-Chloro-5-nitrophenyl)acetonitrile

Purity: 95% CAS: 52427-02-8 MDL Number: MFCD18393747 Molecular Formula: C8H5ClN2O2 Molecular Weight: 196,59 Hazard: Irritant

Formula: C₈H₅ClN₂O₂
Melting Pt: 97…100 °C
MDL Number: MFCD18393747
CAS Number: 52427-02-8

Order Now

Specification Test Results

Melting point 97-100°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR