4-Acetoxythiophene-3-carboxylic acid methyl ester ≥95%
Supplier: Matrix Scientific
058698-1G
058698-5G
101915-464EA
464.62
USD
101915-464
101915-466
4-Acetoxythiophene-3-carboxylic acid methyl ester ≥95%
4-Acetoxythiophene-3-carboxylic acid methyl ester
Purity: 95+%
CAS: 65369-31-5
MDL Number: MFCD11227239
Molecular Formula: C8H8O4S
Molecular Weight: 200,21
Hazard: Irritant
Formula:
C₈H₈O₄S MW: 200.21 g/mol |
MDL Number:
MFCD11227239 CAS Number: 65369-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...