3-Piperazin-1-yl-1,2-benzisoxazole ≥95%
Supplier: Matrix Scientific
058606-250MG
058606-1G
101915-092EA
95.38
USD
101915-092
101915-094
3-Piperazin-1-yl-1,2-benzisoxazole ≥95%
3-Piperazin-1-yl-1,2-benzisoxazole
Purity: 95%
CAS: 87691-89-2
MDL Number: MFCD08060968
Molecular Formula: C11H13N3O
Molecular Weight: 203,25
Hazard: Irritant
Formula:
C₁₁H₁₃N₃O |
MDL Number:
MFCD08060968 CAS Number: 87691-89-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...