N-(2-Carboxyphenyl)glycine

Supplier: Matrix Scientific

Synonyms: 2-((Carboxymethyl)amino)benzoic acid

058470-500MG
101914-872EA 44.62 USD
101914-872
N-(2-Carboxyphenyl)glycine
N-(2-Carboxyphenyl)glycine

Hazard: Irritant

Formula: C₉H₉NO₄
MDL Number: MFCD00020445
CAS Number: 612-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR