Oleic acid cholesteryl ester

Supplier: Matrix Scientific

058399-500MG
101914-732EA 306.15 USD
101914-732
Oleic acid cholesteryl ester
Oleic acid cholesteryl ester

Hazard: Irritant

Formula: C₄₅H₇₈O₂
MDL Number: MFCD00003645
CAS Number: 303-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR