Sudan IV

Supplier: Matrix Scientific
Warning

058388-500MG
101914-708EA 306.15 USD
101914-708
Sudan IV
Sudan IV
Formula: C₂₄H₂₀N₄O
Melting Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00003893
CAS Number: 85-83-6
EINECS: 201-635-8
Merck Index: 12,08538

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR