Ciprofloxacin hydrochloride monohydrate

Supplier: Matrix Scientific

Synonyms: 1-Cyclopropyl- 6-Fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid hydrochloride monohydrate, 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride monohydrate, Nanomycopulitine

058377-500MG
101914-688EA 306.15 USD
101914-688
Ciprofloxacin hydrochloride monohydrate
Ciprofloxacin hydrochloride monohydrate

Hazard: Irritant

Formula: C₁₇H₁₈FN₃O₃·HCl·H₂O
MW: 385.82 g/mol
MDL Number: MFCD00242856
CAS Number: 86393-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR