Atenolol
Supplier: Matrix Scientific
Synonyms:
2-[4-(2-Hydroxy-3-isopropylaminopropoxy)phenyl]acetamide
058293-500MG
101914-524EA
306.15
USD
101914-524
Atenolol
Atenolol
CAS: 29122-68-7
MDL Number: MFCD00057645
Molecular Formula: C14H22N2O3
Molecular Weight: 266,34
Hazard: Irritant
Formula:
C₁₄H₂₂N₂O₃ MW: 266.34 g/mol |
MDL Number:
MFCD00057645 CAS Number: 29122-68-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...