tert-Butyl-4-[(2-amino-5-methylphenyl)amino]piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

058263-500MG 058263-1G
101914-448EA 367.69 USD
101914-448 101914-450
tert-Butyl-4-[(2-amino-5-methylphenyl)amino]piperidine-1-carboxylate ≥95%
tert-Butyl-4-[(2-amino-5-methylphenyl)amino]piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₇H₂₇N₃O₂
MDL Number: MFCD17167310
CAS Number: 950772-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR